Order Entry
Belgium
ContactUsLinkComponent
 
Anticorps monoclonal anti-ERBB2 de lapin [clone : SP3]
  ABCAAB27597-7
 :  Abcam
Anticorps monoclonal anti-ERBB2 de lapin [clone : SP3]
  ABCAAB27597-7
 :  Abcam

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
  • Symbole de l'antigène:
    ERBB2
  • Clonalité:
    Monoclonal
  • Clone:
    SP3
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Cdt:
    7 mL

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type : Primaire
Antigène : ERBB2 (homologue 2 de l'oncogène viral de la leucémie érythroblastique aviaire, v-erb-b2)
Clonalité : Monoclonal
Clone : SP3
Conjugaison: Non conjugué
Epitope :
Hôte : Lapin
Isotype: IgG
Réactivité :