Anticorps polyclonal de lapin anti-AGE
ABCAAB23722-200
: Abcam
- Type d'anticorps:Primaire
- Nom de l'antigène:Advanced Glycosylation End Product
- Symbole de l'antigène:AGE
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Température de stockage:Cool
- Température de transport:Ice
- Taille:200 µl
- Type de conditionnement:Vial
- Cdt:200 µl
:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type : Primaire
Antigène : AGE
Clonalité : Polyclonal
Clone:
Conjugaison : Non conjugué
Epitope :
Hôte : Lapin
Isotype: IgG
Réactivité : Humain, rat