Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:BH3 interacting domain death agonist
- Symbole de l'antigène:BID
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Western blot:Yes
- Température de stockage:Cool
- Température de transport:Ice
- Taille:50 µl
- Type de conditionnement:Vial
- Cdt:50 µl
Spécifications
À propos de cet article
Rabbit polyclonal to Bid Cleavage Site.
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type : Primaire
Antigène : BID (agoniste de la mort du domaine d'interaction BH3)
Clonalité : Polyclonal
Clone:
Conjugaison : Non conjugué
Epitope :
Hôte : Lapin
Isotype : aucun
Réactivité : Souris