Order Entry
Belgium
ContactUsLinkComponent
Anti-Insulin Receptor (phospho Y972) Rabbit Polyclonal Antibody
Anti-Insulin Receptor (phospho Y972) Rabbit Polyclonal Antibody
Catalog # ABCAAB5678-50
Supplier:  Abcam
undefined
Anti-Insulin Receptor (phospho Y972) Rabbit Polyclonal Antibody
Catalog # ABCAAB5678-50
Supplier:  Abcam

Specifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    insulin receptor
  • Symbole de l'antigène:
    INSR
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • Précipitation immunologique:
    Yes
  • Western blot:
    Yes
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Taille:
    50 µl
  • Type de conditionnement:
    Vial
  • Cdt:
    50 µl

Specifications

About this item

Rabbit polyclonal to Insulin Receptor (phospho Y972).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Insulin Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human