Anticorps polyclonal de lapin anti-PTK2B
ABCAAB4807-50
: Abcam
- Type d'anticorps:Primaire
- Nom de l'antigène:Protein Tyrosine Kinase 2 beta
- Symbole de l'antigène:PTK2B
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Western blot:Yes
- Température de stockage:Cool
- Température de transport:Ice
- Taille:50 µl
- Type de conditionnement:Vial
- Cdt:50 µl
Rabbit polyclonal to PYK2 (phospho Y579 + Y580).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type : Primaire
Antigène : PTK2B (protéine tyrosine kinase 2 bêta)
Clonalité : Polyclonal
Clone:
Conjugaison : Non conjugué
Epitope :
Hôte : Lapin
Isotype: IgG
Réactivité :