Order Entry
Belgium
ContactUsLinkComponent
 
Anti-TRAIL Mouse Monoclonal Antibody [clone: 200000]
  ABCAAB2219-100
 :  Abcam
undefined
Anti-TRAIL Mouse Monoclonal Antibody [clone: 200000]
  ABCAAB2219-100
 :  Abcam

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    tumor necrosis factor (ligand) superfamily, member 10
  • Symbole de l'antigène:
    TNFSF10
  • Clonalité:
    Monoclonal
  • Clone:
    2E5
  • Conjugaison:
    Unconjugated
  • Cytométrie en flux:
    Yes
  • Hôte:
    Mouse
  • Seq:
    1286
  • Neutralisation:
    Yes
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Taille:
    100 µg
  • Type de conditionnement:
    Vial
  • Cdt:
    100 µG

 

 

Mouse monoclonal [2E5] to TRAIL.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type : Primaire
Antigène : TNFSF10 (superfamille du facteur (ligand) de nécrose tumorale, membre 10)
Clonalité : Monoclonal
Clone: 200000
Conjugaison : Non conjugué
Epitope :
Hôte : Souris
Isotype: IgG1
Réactivité: Humain