Anti-L type Calcium CP gamma 2 Rabbit Polyclonal Antibody
BIRBORB41220-100
: Biorbyt
- Type d'anticorps:Primaire
- Nom de l'antigène:L type Calcium CP gamma 2
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Nombre d'acides aminés:93 to 311
- Storage buffer:0,1M Tris, 0,1M Glycine, 20% Glycerol (pH7).
- Température de stockage:Cool
- Concentration:0,58 mg/ml
- Température de transport:Ice
- Immunogène:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 93 and 311 of Human CACNG2.
- Purification:purified by antigen affinity chromatography
- Taille:100 µl
- Cdt:100 µl
Western Blot: 1:500-1:3000
Type: Primary
Antigen: L type Calcium CP gamma 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human