Anti-CDCA7 Rabbit Polyclonal Antibody
N° de catalogue ORIGTA315887
Fournisseur: OriGene
Aller à :
Product Details & Documents
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000
Type: Primary
Antigen: CDCA7 (cell division cycle associated 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
TA315887.pdf
Fiche technique
Spécifications
- Western blot:Yes
- Adsorption croisée:none
- Type d'anticorps:Primaire
- Formulaire:Liquid
- Purification:purified by affinity chromatography
- Symbole de l'antigène:CDCA7
- Isotype:IgG
- Conjugaison:Unconjugated
- Réactivité:Human|Rat|Mouse
- Température de stockage:Cool
- Cat. No.:ORIGTA315887
- Clonalité:Polyclonal
- Épitope:internal
- Nom de l'antigène:Cell division cycle associated 7
- Hôte:Rabbit
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Température de transport:Ice
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



