Order Entry
Belgium
ContactUsLinkComponent
Anti-CDK5R1 Rabbit Polyclonal Antibody
Anti-CDK5R1 Rabbit Polyclonal Antibody
Catalog # ORIGTA314819
Supplier:  OriGene
Anti-CDK5R1 Rabbit Polyclonal Antibody
Catalog # ORIGTA314819
Supplier:  OriGene

Specifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Cyclin-dependent kinase 5 regulatory subunit 1
  • Symbole de l'antigène:
    CDK5R1
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Épitope:
    internal
  • Adsorption croisée:
    none
  • Formulaire:
    Liquid
  • Storage buffer:
    Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Purification:
    purified by affinity chromatography
  • Cdt:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: CDK5R1 (cyclin-dependent kinase 5, regulatory subunit 1 (p35))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat