Anti-RPL34 Rabbit Polyclonal Antibody
Catalog # ORIGTA314468
Supplier: OriGene
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Ribosomal Protein L34
- Symbole de l'antigène:RPL34
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Environmentally Preferable:
- Épitope:internal
- Adsorption croisée:none
- Formulaire:Liquid
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by affinity chromatography
- Cdt:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000
Type: Primary
Antigen: RPL34 (ribosomal protein L34)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat