Anti-IRAK1 Rabbit Polyclonal Antibody
Catalog # ORIGTA314086
Supplier: OriGene
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:interleukin-1 receptor-associated kinase 1
- Symbole de l'antigène:IRAK1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human
- Épitope:internal
- Adsorption croisée:none
- Formulaire:Liquid
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by affinity chromatography
- Cdt:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:5000
Type: Primary
Antigen: IRAK1 (interleukin-1 receptor-associated kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human