Anti-EGR3 Rabbit Polyclonal Antibody
ORIGTA313796
: OriGene
- Type d'anticorps:Primaire
- Nom de l'antigène:early growth response 3
- Symbole de l'antigène:EGR3
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Épitope:internal
- Adsorption croisée:none
- Formulaire:Liquid
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by affinity chromatography
- Cdt:100 µl
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000
Type: Primary
Antigen: EGR3 (early growth response 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat