Order Entry
Belgium
ContactUsLinkComponent
 
Anti-ATP5G3 Rabbit Polyclonal Antibody
  ORIGTA313490
 :  OriGene
Anti-ATP5G3 Rabbit Polyclonal Antibody
  ORIGTA313490
 :  OriGene

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C3 (subunit 9)
  • Symbole de l'antigène:
    ATP5G3
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Épitope:
    internal
  • Adsorption croisée:
    none
  • Formulaire:
    Liquid
  • Storage buffer:
    Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Purification:
    purified by affinity chromatography
  • Cdt:
    100 µl

 

 

Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: ATP5G3 (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C3 (subunit 9))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat