Order Entry
Belgium
ContactUsLinkComponent
Anti-PURG Rabbit Polyclonal Antibody
Anti-PURG Rabbit Polyclonal Antibody
Catalog # AVIVARP50381_P050
Anti-PURG Rabbit Polyclonal Antibody
Catalog # AVIVARP50381_P050

Specifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Purine-rich element binding protein G
  • Symbole de l'antigène:
    PURG
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • Western blot:
    Yes
  • Épitope:
    N-terminal
  • Adsorption croisée:
    none
  • ID de gène:
    29942
  • Synonymes antigène:
    PURG-B|PURG-A
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Cdt:
    50 µG

Specifications

About this item

Type: Primary
Antigen: PURG (purine-rich element binding protein G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: