Adapters with screw thread and standard gound joint cone
Catalog # QVFLVBS0101404
Supplier: QVF LABORTECHNIK
Specifications
- Rodage mâle:29/32 NS
- Pour:
- Ø trou:8,8 - 9,3 mm
- Filetage:GL 18
- Type:
- Environmentally Preferable:
- Cdt:1
Specifications
About this item
Connecters, ground cone with thread and screw cap, can be used to assemble the thermometers without ground joint. The seal is part of delivery.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes