Permanent marker pen, Edding 3000
N° de catalogue 811-3123
Fournisseur: THEODOR KIHLER BURO EINRICHTUN
Spécifications
- Couleur:bleu
- Désignation:Permanent marker, wedge tip, Edding 3000
- Cdt:1
Spécifications
À propos de cet article
Permananent marker for writing, colouring, labelling for materials such as metal, glass or plastic, low odour ink, round interchangeable tip. Line thickness of 1,5 - 3 mm.
- Fast drying
- Waterproof
- Refillable