Chromium test kits, models CH-8 and CH-12, comparator method
Catalog # HACH183400
Supplier: Hach
CAS Number:
Specifications
- Méthode chimique:Diphenylcarbazide hypobromite oxidation
- Cdt:1 KIT
- Désignation:Chromium (VI) test kit, model CH-8, colour disc
- Plage de mesure:0 - 1,3 mg/l
Specifications
About this item
These kits are used to determine chromium or chromium(VI) using simple, economical colour disc methods.
Delivery: CH-12 kit includes four reagents, clippers, two colour discs, comparator box, two viewing tubes, bottle, flask, dropper tube, stove and cover, test tube clamp, instructions and carrying case. CH-8 kit includes one reagent, two colour discs, comparator box, two viewing tubes, vial, instructions and carrying case.