Trolley for gas cylinders
N° de catalogue MSGA13267
Fournisseur: asecos
Spécifications
- Désignation:Trolley for carrying gas cylinders up to 4 (Ø 230 mm)
- Environmentally Preferable:
- Cdt:1
Spécifications
À propos de cet article
This trolley made of PE can equip up to 4 cylinders with a load capacity of 300 kg. Safe transport is ensured with the provision for forklift.
Livraison: With lashing straps and safety chain.