ContactUsLinkComponent
Agarose Sealing Solution
  786-226
undefined
Agarose Sealing Solution
  786-226
  • Désignation:
    Agarose Sealing Solution
  • Environmentally Preferable:
  • Cdt:
    50 mL
Simply heat and use for sealing the IPG strips for SDS-PAGE analysis. The Agarose Sealing Solution is prepared in a proprietary buffer to minimise reoxidation of the competing thiol pairs as proteins enter into the second dimension gel. Improves resolution and prevents streaking of protein spots on 2D gels.