Draining racks, wall mounted
Draining rack, with 16 rods of 200 mm and 16 rods of 140 mm, 450×600 mm
N° de catalogue SEMA2110
Fournisseur: SEMADENI
Aller à :
Product Details & Documents
White, PVC drain channel, polyethylene coated metal rack.
- Drain channels inclined toward the centre and provided with a drain point
- Metal frame and drip plasticised PE rods
- The rack is hung on the wall plate and can be easily removed for cleaning
Spécifications
- Désignation:Draining rack, with 16 rods of 200 mm and 16 rods of 140 mm
- Seq:02
- l×h:450×600 mm