Stopcocks, one-way
Stopcocks, one-way
Female Luer to Female Luer
N° de catalogue POPR6020
Fournisseur:   CADENCE SCIENCE
Stopcocks, one-way
N° de catalogue POPR6020
Fournisseur:  CADENCE SCIENCE
Stopcocks, one-way
Female Luer to Female Luer
Prix
321,00 €
Connexion Pour la disponibilité
1 * 1 Pce
Product Details & Documents
Theses stopcocks, made of nickel plated brass, are precision-machined, individually lapped, and carefully mated to maintain pressure characteristics. These stopcocks are easily integrated with other stopcocks, manifolds, adapters, tubing, needles, and glass syringes.
Spécifications
  • Désignation:
    Female Luer to Female Luer
  • Seq:
    02