Protein solubilisation buffer
Protein solubilisation buffer
FOCUS™ protein solubilisation buffer (PSB) with Diluent III, 50 mL
N° de catalogue 786-PSB
Fournisseur:   G-Biosciences
Protein solubilisation buffer
N° de catalogue 786-PSB
Fournisseur:  G-Biosciences
Numéro CAS:  
Protein solubilisation buffer
Protein solubilisation buffer
FOCUS™ protein solubilisation buffer (PSB) with Diluent III, 50 mL
Numéro CAS:  
Prix
213,00 €
Connexion Pour la disponibilité
1 * 50 mL
Product Details & Documents
FOCUS™ protein solubilisation buffer is a dry, urea-based buffer for protein solubilisation. It is a pre-mixed formulation of urea, thiourea, CHAPS and non detergent sulphobetaine for maximum solubilising strength.
  • Convenient and simple to use - simply hydrate and use
  • Prevents urea induced protein carbamylation
  • Prevents waste of unused reagents
Livraison: Supplied with specific dilution buffer for optimal rehydration and protein solubilisation conditions.
Documents
Spécifications
Spécifications
  • Cdt:
    50 mL
  • Désignation:
    FOCUS™ protein solubilisation buffer (PSB) with Diluent III