Counting chamber, McMaster
Counting chamber, McMaster, 1,5 mm
N° de catalogue HAWKZ11000
Fournisseur: HAWKSLEY AND SONS
Aller à :
Product Details & Documents
Counting chamber with cover glass and two counting areas.
- 10×10 mm grid etched on the under side of the top cover
Suitable for worm egg counts.
Spécifications
- Profondeur de chambre:1,5 mm
- Type:McMaster
- Désignation:Counting chamber