Microbact™
Microbact™ GNB 24E, 40 Tests
N° de catalogue OXOIMB1131A
Microbact™
N° de catalogue OXOIMB1131A
Numéro CAS:  
Microbact™
Microbact™ GNB 24E, 40 Tests
Numéro CAS:  
Prix
344,00 €
Connexion Pour la disponibilité
1 * 40 Tests
Product Details & Documents
The Microbact™ Gram-negative system is to be used for the identification of aerobic and facultatively anaerobic Gram-negative bacteria (Enterobacteriaceae and miscellaneous Gram- negative bacteria). Organism identification is based on pH change and substrate utilisations as established by published reference methodologies.
  • Fast
  • Simple
  • Reliable
Spécifications
Spécifications
  • Cdt:
    40 Tests
  • Désignation:
    Microbact™ GNB 24E