Cell surface protein isolation and labelling kit, HOOK™
Cell surface protein isolation and labelling kit, HOOK™
HOOK™ Cell Surface Protein Isolation and Labeling Kit, complete, 1 KIT
N° de catalogue 786-316
Fournisseur:   G-Biosciences
Cell surface protein isolation and labelling kit, HOOK™
N° de catalogue 786-316
Fournisseur:  G-Biosciences
Numéro CAS:  
Cell surface protein isolation and labelling kit, HOOK™
Cell surface protein isolation and labelling kit, HOOK™
HOOK™ Cell Surface Protein Isolation and Labeling Kit, complete, 1 KIT
Numéro CAS:  
Prix
411,00 €
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
The HOOK™ Cell Surface Protein Isolation Kit uses HOOK™ biotin labelling and purification technology in conjunction with Mammalian Cell PE LB™ Lysis Buffer to conveniently label cell surface proteins and isolate them for further analysis, including Western blotting. The Kit is supplied with all the necessary reagents and buffers for convenience and improved reproducibility. The kit is compatible with a wide variety of mammalian cells and can be used to compare treated and untreated cells and differences between different cell lines.
Informations de commande: Accessories included. Additional supplies available on request, please contact your local Avantor sales office for further details.
Documents
Spécifications
Spécifications
  • Cdt:
    1 KIT
  • Désignation:
    HOOK™ Cell Surface Protein Isolation and Labeling Kit, complete