Deep well plates, 96-well, Bioblock™
730-1091
- Emballage:4 plaques/sachet
- Volume de travail recommandé:1,0 ml
- Couleur:naturel
- Désignation:Plaque à 96 puits profonds, PS, 1,2 ml
- l×p×h:127,8×85,5×41,6 mm
- Cdt:24
- Volume puits:1,2 ml
PS. The 1,2 ml capacity round bottom deep well plate (1 ml when capped) can withstand centrifugation up to 3000×g. It is not DMSO resistant.
: Packed in bags of 4 plates.