Silicone paste BAYER assortment, KORASILON®
Silicone paste BAYER assortment, KORASILON®
Silicone paste, KORASILON®
N° de catalogue ROTH0858.1
Marketsource
Fournisseur:   Roth Carl
Silicone paste BAYER assortment, KORASILON®
N° de catalogue ROTH0858.1
Fournisseur:  Roth Carl
Marketsource
Silicone paste BAYER assortment, KORASILON®
Silicone paste, KORASILON®
Prix
30,50 €
Connexion Pour la disponibilité
1 * 6 ST
Product Details & Documents
Assortment of 2 × each tube containing low viscosity, medium viscosity and high viscosity silicone paste.
Spécifications
  • Désignation:
    Silicone paste, KORASILON®