Analytical HPLC columns, XBridge™ BEH C8
Fournisseur: WATERS
Certificats
À propos de cet article
The XBridge™ BEH C8 sorbent is less retentive compared to the XBridge™ BEH C18 sorbent. The trifunctionally bonded sorbent offers the widest usable pH range (1 to 12), superior low pH stability and ultra-low column bleed.
Spécifications
- Catalog No:
- WATE186003055
- WATE186003054
- WATE186003192
- Charge de carbone:
- 13%
- 13%
- 13%
- End-capping:
- Oui
- Oui
- Oui
- Plage de pH:
- 1 - 12
- 1 - 12
- 1 - 12
- Surface area:
- 185 m²/g
- 185 m²/g
- 185 m²/g
- Code USP:
- L7
- L7
- L7