Tissue forceps, Aesculap® SUSI®
Tissue forceps, Aesculap® SUSI®
Tissue forceps 1:2, 145 mm
N° de catalogue AESCBD557SU
Fournisseur:   Aesculap
Tissue forceps, Aesculap® SUSI®
N° de catalogue AESCBD557SU
Fournisseur:  Aesculap
Tissue forceps, Aesculap® SUSI®
Tissue forceps 1:2, 145 mm
Prix
70,30 €
Connexion Pour la disponibilité
1 * 20 ST
Product Details & Documents
These disposable forceps are made from a high performance polymer. The special composition of this high-grade polymer ensures an outstanding quality and provides optimal mechanical properties.
  • Material ensures a safe, easy and environmentally friendly removal through incineration in the regular hospital waste removal system
  • Single use: Reduces the risk of cross-contamination
  • 1×2 teeth
  • Sterile single packed in dispenser box
Spécifications
  • Type:
    Tissue forceps 1:2
  • Longueur:
    145 mm