Pasteur pipettes, disposable, LDPE
7,3 ml, 300 mm, - ml
N° de catalogue SEMA4208
Fournisseur: SEMADENI
Aller à :
Product Details & Documents
LDPE, disposable, non-sterile.
- Almost unbreakable transfer pipettes
- Good chemical resistance and can be used at low temperatures
- Pipettes are made from one piece: No risk of leakage
- Dustproof packaged in convenient standard boxes
Spécifications
- Longueur:300 mm
- Seq:04
- Division échelle:- ml
- Volume d'aspiration:7,3 ml