Versene
Catalog # L0630-100
Supplier: Biowest
CAS Number:
Specifications
- Cdt:100 ml
- Désignation:Versene
Specifications
About this item
Versene can be used instead of trypsin. It is a chelating agent that disperses the cells by cutting the cytoplasmic bridges between them. Versene has the advantage of being able to withstand autoclave temperature, which is a guarantee of sterility. In addition, it is less aggressive cells than trypsin, which is useful for studies on cell growth.