Sample cups for analyser
For Coulter Counter analysers, with cap, 20 ml, Bag of 250 or 1250/case
Catalog # 391-3720
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
Cups made of PS.
- Conical bottom and supported with skirts
- Good chemical resistance
- Good temperature resistance
Suitable for analytical systems, such as Technicon, Gemsaec, Centrifichem, Hitachi, TOA and Coulter analysers.
Specifications
- Emballage:Bag of 250 or 1250/case
- Capacité:20 ml
- Désignation:For Coulter Counter analysers, with cap
- Seq:05