Microscope slide, UniMark®
Micoscope slides, UniMark
N° de catalogue MARI0704022
Fournisseur: MARIENFELD
Aller à :
Product Details & Documents
UniMark slide is designed with a white circle of Ø 12 mm and a white marking area of 20 mm for use in cytocentrifuges.
It is suitable for suitable for marking with inkjet of laser printers and permanent markers.
Certifications: DIN 8037/1. IVD 98/79/EC.
Emballage: 50 pieces in a box.
Spécifications
- Désignation:Micoscope slides, UniMark