Solid phase extraction cartridges, Oasis® HLB
Solid phase extraction cartridges, Oasis® HLB
Marketsource
Fournisseur:   WATERS
Solid phase extraction cartridges, Oasis® HLB
Fournisseur:  WATERS
Marketsource
Certificats
Solid phase extraction cartridges, Oasis® HLB
3 Options disponibles ci-dessous
Product Details & Documents
Oasis® HLB cartridges contain the Oasis® HLB sorbent, which is a universal polymeric reversed phase sorbent that was developed for the extraction of a wide range of acidic, basic, and neutral compounds from various matrices using a simple, generic protocol.
Since the HLB sorbent is water wettable, it maintains its capability for higher retention and excellent recoveries even if the sorbent runs dry, which means there is no need to take extraordinary steps to keep the sorbent beds from drying out during the critical steps prior to sample loading.
These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.
Product Family Options
Informations sur le produit
  • Désignation
    Type de colonne
    Taille de pore
    Granulométrie
    Capacité
    Poids du lit
    Disponibilité
    Prix
  • Oasis® HLB 6 cc Vac cartridge
    Cartouche
    80 Å
    60 µm
    6 ml
    500 mg
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 30 ST
    1 * 30 ST - 308,00 €
    308,00 €
    1 * 30 ST
       308,00 €
    Spécifications :
    Marketsource
    Marketsource
  • Oasis® HLB 20 cc Vac cartridge
    Cartouche
    80 Å
    60 µm
    20 ml
    1000 mg
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 20 ST
    1 * 20 ST - 403,00 €
    403,00 €
    1 * 20 ST
       403,00 €
    Spécifications :
    Marketsource
    Marketsource
  • Oasis® HLB 6 cc Vac cartridge
    Cartouche
    80 Å
    30 µm
    6 ml
    200 mg
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 30 ST
    1 * 30 ST - 214,00 €
    214,00 €
    1 * 30 ST
       214,00 €
    Spécifications :
    Marketsource
    Marketsource