Micro scissors for endoscopic work, Aesculap® PaediScope®
Micro scissors for intraventricular neuroendoscopy, flexible, 250 mm
N° de catalogue AESCFF373R
Fournisseur: Aesculap
Micro scissors for endoscopic work, Aesculap® PaediScope®
N° de catalogue AESCFF373R
Fournisseur: Aesculap
Aller à :
Product Details & Documents
Flexible micro scissors for advanced intraventricular Neuroendoscopy.
Ø = 1 mm
Spécifications
- Type:Micro scissors for intraventricular neuroendoscopy, flexible
- Longueur:250 mm
- Seq:01