ContactUsLinkComponent
Deuterium lamp, TruLife™ 1,5
Deuterium lamp, TruLife™ 1,5
Catalog # PERCL6022728
Supplier:  PerkinElmer
Deuterium lamp, TruLife™ 1,5
Catalog # PERCL6022728
Supplier:  PerkinElmer
Specifications
  • Désignation:
    TruLife™ 1.5 UV deuterium lamp
  • Seq:
    02
  • Cdt:
    1
Specifications
About this item
These deuterium lamps provide the best wavelength coverage across the working range of the spectrometer for consistently high performance.
  • Lamps are pre-aligned for easy installation
  • Extended lifetime of 1500 hours
  • Built-in digital timer to monitor lamp lifetime