ContactUsLinkComponent
Cell culture media, Schneider's Insect Medium
Cell culture media, Schneider's Insect Medium
N° de catalogue L0207-500
Fournisseur:  Biowest
Numéro CAS:  
undefined
Cell culture media, Schneider's Insect Medium
N° de catalogue L0207-500
Fournisseur:  Biowest
Numéro CAS:  
Spécifications
  • Format du milieu:
    Liquid
  • Cdt:
    500 mL
  • Désignation:
    Milieu insecte de Schneider (liquide)
Spécifications
À propos de cet article
Schneider’s Insect Medium was developed to support the growth of excised imaginal discs from the fruit fly, Drosophila melanogaster. When supplemented with 5 to 20% heat-inactivated foetal bovine serum Schneider's mediuam has been found to support the rapid growth of both primary and established cultures of cells derived from Drosophila spp. and several other dipterans.