Examination table drapes
Examination table drapes, pre-cut (180 sheets), bleu, 50×38 cm
N° de catalogue LCHMDRP-01A
Fournisseur: LCH MEDICAL
Aller à :
Product Details & Documents
Protective drape for examination tables. This latex-free cellulose cotton wool/transparent embossed polyethylene drape can be used as protective drape, napkin, bib, thanks to its excellent protection and absorption.
- Two layers: Cellulose cotton wool and transparent polyethylene
- High absorption features
- Plastified, embossed, impermeable
Pre-cut: 380 mm
Spécifications
- Couleur:bleu
- Désignation:Examination table drapes, pre-cut (180 sheets)
- L×l:50×38 cm



