Quick-connect coupling
For connecting and disconneting hoses and devices, with 1/8“ hose barb and shutoff, 3,2 mm
Catalog # 229-5123
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Jump to:
Product Details & Documents
Quick-connect coupling and plug connection, made of acetal (POM) for a complete connection.
- High mechanical stability
- Extremely fatigue strength
- Good electrical insulating features
- Excellent chemical resistance
- Both parts are with barrier
Specifications
- Tube Ø int.:3,2 mm
- Désignation:For connecting and disconneting hoses and devices, with 1/8“ hose barb and shutoff