Closed caps, PTFE lined
PTFE faced silicone liner, 24-414, blanc
N° de catalogue EPSCB267-2414UBB
Fournisseur: EP SCIENTIFIC
Closed caps, PTFE lined
N° de catalogue EPSCB267-2414UBB
Fournisseur: EP SCIENTIFIC
Aller à :
Product Details & Documents
These PTFE-lined closed caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials.
The PTFE liner is bonded to the cap so it won't fall out or become separated from the cap.
Spécifications
- Filetage:24-414
- Couleur:blanc
- Désignation:PTFE faced silicone liner
- Seq:01
- Cat. No.:EPSCB267-2414UBB
- Certification environnementale:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact