À propos de cet article
DyNAmo™ qPCR Kits offer excellent performance in detection and quantitation of DNA and RNA sequences from various sources. The DyNAmo™ qPCR kit family features optimised kits for fast and conventional qPCR. Kits utilising either SYBR® Green or probe chemistries for various platforms are available. All DyNAmo™ qPCR kits are provided as convenient 2X master mixes - only template and primers need to be added.
- Extremely fast protocols: combined annealing and extension step of only 15 seconds
- Specific and sensitive detection of a wide range of template concentration
- Included dUTP allows the use of UNG for prevention of carry-over contamination
- Convenient 2X master mix and optimised protocols
- Validated with all major real time instruments
- Licenced for qPCR
DyNAmo™ Flash SYBR® Green qPCR Kit and DyNAmo™ Flash Probe qPCR Kit offer enhanced performance for real time qPCR. These kits provide sensitive and reproducible detection of target DNA with shorter run times in both fast and conventional instruments.
DyNAmo™ Flash SYBR® Green qPCR Kit provides qPCR results faster than other SYBR® Green kits without compromising the qPCR performance. A DNA-binding domain attached to the polymerase in this kit improves the physical stability of the polymerase-DNA complex, thus enhancing the processivity and robustness of the amplification. The high amplification efficiency of DyNAmo™ Flash SYBR® Green qPCR Kit gives reliable quantitation and early C(t) values. Due to the high signal intensity, DyNAmo™ Flash SYBR® Green qPCR provides an excellent signal-to-noise ratio.
Spécifications
- Application:qPCR dye