Ladies sweat jacket, with hood, Acode® Basecamp, 1746
KWIN100234940M
- Taille de confection:M
- Type de fermeture:Avec fermeture éclair
- Applications des tenues:À usage universel
- Genre:Pour femmes
- Zone à risque:Froid
- Couleur:noir
- Seq:06
- Modèle:1746
- Tissu:66% cotton and 34% polyester
- Longueur de manches ou de jambes:Long
- Poids:320 g/m²
- Cdt:1
Double face sweatshirt made of 66% cotton and 34% polyester.
- Combed cotton
- Inside of hood, zipper and seams in contrasting colour
- Tailored fit
- Front pockets
- Rib knitted cuffs with elastane
Weight: 320 g/m²