DALT Buffer kit for precast gels
DALT Buffer kit for precast gels
DALT buffer kit for 12 precast gels, 1 KIT
N° de catalogue 17-6002-50
Fournisseur:   Cytiva
DALT Buffer kit for precast gels
N° de catalogue 17-6002-50
Fournisseur:  Cytiva
Numéro CAS:  
DALT Buffer kit for precast gels
DALT buffer kit for 12 precast gels, 1 KIT
Numéro CAS:  
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
DALT Buffer kit is used together with DALT Gel 12,5, which comprises concentrated buffers for running the gel, gel buffer for inserting the gel into DALT precast gel cassette and sealing solution for application of the IPG strip. The buffer in the gel is part of a buffer system that gives longer shelf-life.
Buffer based on piperidinopropionamide (PPA)
Spécifications
Spécifications
  • Cdt:
    1 KIT
  • Désignation:
    DALT buffer kit for 12 precast gels
Spécifications