SHEL-LAB-SMOHP955-590.jpg
SHEL LAB SMOHP High-Performance ovens
SHEL LAB SMOHP High-Performance Ovens | Avantor
SHEL LAB High-performance ovens have rugged, welded construction with a double-walled, corrosion-resistant stainless steel interior and a powder-coated exterior. They are engineered for safe, accurate and uniform performance in demanding laboratory environments.
  • Control, timer program and monitoring
  • Integrated watlow control panel
  • Fast ramp-up to 306 °C* for faster cycle times (*260 °C for SMO14HP-2 and SMO38HP-2)
  • Stainless steel interior construction for long life operation, easy cleaning
  • Double-wall construction to minimize external surface temperature
  • High-Performance forced-air circulation to assure maximum temperature uniformity
  • Built-in overtemperature protection
  • Temperature stability: 0.3 °C
These ovens have their applications in Asphalt testing, ASTM, UL and life testing, baking, conditioning, curing, moisture and stability testing, drying, burn-in and aging.
Horizontal airflow ensures uniform heating profiles and powerful heating elements provide fast heat-up and recovery to ensure rapid drying. Heated air is continuously circulated by twin turbo blowers. Blowers also circulate air around motor bearings to prolong motor life.
Punched stainless steel shelves are height-adjustable. Adjustable three-inch diameter air intake and exhaust ports can be opened for fast drying of high-moisture content samples.
SHEL-LAB-SMO5HP400-495.jpg
SHEL LAB High-Performance Ovens are engineered to meet the most critical temperature requirements.
SHEL-LAB-SMO10HP400-495.jpg
SHEL LAB High-Performance Ovens are engineered to meet the most critical temperature requirements.
SHEL-LAB-SMO28HP400-495.jpg
SHEL LAB High-Performance Ovens are engineered to meet the most critical temperature requirements.