Single-Use samplers, for the highest standards in sampling
Cleaning of samplers is a demanding, time-intensive task, especially when the material is greasy, strongly adhesive or toxic. Single-use is the solution
Sampling lances - for liquids
Single-use samplers for thin liquids to viscous fluids.
Disposable dippers and ladles
Long handled ladles for sampling of liquids and viscous media, the DispoDippers furthermore combine sampler and sampling container in one device.
Sampling lances - for powders and granules
Single-use samplers for various applications like sampling of small amounts of powders to sampling of larger quantities of small grain granulates.
Scoops, scrapers, spoons and spatulas
Sampling tools to collect, handle and process sampling materials from very small (0,5 ml) to larger volumes (2 l)
Detectable sampling tools
Sampling tools that have a special additive that makes them visible to colour scanners, metal detectors or X rays, even though they are made of PS.
Blue colour sampling tools for increased visibility
Blue products, or parts of them, can be quickly found and easily identified because there is no natural food product that has this colour.
- Food sampling scoops, blue
- Sampling Scoops with long handle, blue
- Food Sampling scrapers, blue
- Food Sampling spoons, blue
- Sampling Scoops with long handle, blue
- Sampling Spoons with long handle, curved, blue
- Dosing spoon, blue
- Food Sampling spatulas, blue
- Sampling palette knife spatula, blue
- Forceps, blue, angled/pointed, pointed or with broad tip
- Ladle, blue, with rounded corners and long handle
Bürkle Bio line - the purity of disposable products and ecological sustainability combined perfectly
Made from renewable and 100% recyclable raw materials these products help to combine highest quality demands on sampling tools and sustainability
Disposable forceps for precise lab work
Ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples.