Orders LinkContactUsLinkComponent
biodegradability1920x760.jpg
OxiTop - Easy way to manometric bio-degradation
Webinar
Thursday, 4th October, 2022
10:00 to 11:00 CET (Berlin, Paris, Madrid)
Replay
OxiTop - Easy way to manometric bio-degradation | Avantor
WebinarEmailAlerts.jpg
Email alerts
WebinarFreeRegistration.jpg
Free registration
Webinar-chat-area.jpg
Chat area
webinar-Certificate-of-attendance.jpg
Certificate of attendance
Respirometric measurements like with OxiTop®, have been a well-established term in the measurement of Biochemical Oxygen Demand (BOD) for self-monitoring and advanced manometric respiration testing for decades, especially in municipal wastewater treatment plants.
In this webinar you will receive extensive information about the different methods and equipment to determine
  • Dilution BOD
  • Self-monitoring measurement of BOD in undiluted samples according to DIN EN 1899-2
  • OECD biodegradability tests (OECD/microbiology – lab procedure according to DIN ISO 29 408 / ISO 9408 / OECD 301 F)
  • Soil respiration (OECD/microbiology – lab procedure according to DIN ISO 29 408 / ISO 9408 / OECD 301 F)
  • Biogas testing and more
For more information, please contact us at [email protected]
Presented by:
DrKlausReithmayer100x100.jpg
Dr. Klaus Reithmayer
Dr. Klaus Reithmayer Born and living in Munich, married, two adult daughters, hobbies: playing classical guitar, horse riding. Study of mineralogy (diploma), thesis in crystallography, both at Ludwigs-Maximilians university in Munich, Bavaria. Various positions in sales, since 1999 in the laboratory business, since 2001 Product Manager at WTW, today Senior Product Manager for lab and field electrochemistry and OxiTop® at Xylem Analytics Germany Sales GmbH & Co.KG.