Orders LinkContactUsLinkComponent
Flasks | Avantor
Flasks
From education to research and more, science requires a variety of flasks. Avantor® offers an extensive selection of laboratory flasks for every application.
Disposable or autoclavable models with flat or baffled bases in a variety of graduation ranges and capacities.
Products feature vented, plug-seal or phenolic style caps for tight closure.
Conical Flasks
Mix and store liquids used in chemistry labs. Available in a variety of capacities to hold any sample size required.
A wide variety of flasks to meet your laboratory needs.
Choose a single heavy-duty neck or up to four angled or vertical necks to suit your purpose.
Autoclavable flasks of varying capacities and graduation ranges are available, including microscale options.
Borosilicate glass models feature heavy wall construction in an array of capacities.
Conical glass flasks with glass or PTFE stoppers feature wide collars that serve as funnels.
Borosilicate glass construction and controlled wall thickness provide maximum chemical resistance and shock protection.
Round and pear-shaped models of borosilicate glass with vertical or angled side arms for storage, or to perform simple reactions.
Borosilicate glass products are offered with heavy wall construction. May be uncoated or polycoated for durability.
Made of borosilicate glass in an array of capacities, including large-scale options.
A wide offering for distillation, chemical reactions, heating liquid samples and storage.
Borosilicate glass and disposable polystyrene flasks for the growth of cell lines using microcarrier beads.
Prepare and hold highly volatile substances when conducting electrophilic aromatic substitutions.