Orders LinkContactUsLinkComponent
Image of an Avantor bioprocess system in the foreground, with two scientists standing in the background
Scalable Systems Tailored to Your Needs
Custom bioprocess systems meet pipeline, regulatory, and capacity needs without sacrificing performance. Avantor is your one-stop shop delivering engineer-to-order (ETO) and configure-to-order (CTO) bioprocess solutions including mixing, buffer preparation, TFF, all supported by single-use consumables and CFR Part 11–compliant automation.
Consult a Specialist
Custom Bioprocess Systems | Avantor
Image of an Avantor mixing system
Scalable Mixing with Low Shear Control
Magnetic mixers from 500mL to 2500L support gentle, high-performance blending for liquid-liquid or solids-liquid mixing with configure- and engineer-to-order flexibility.
Image of an Avantor tangential flow filtration system
TFF Systems for Purification
Engineer fully automated TFF systems for hollow fiber or cassette filters, designed to support crossflow filtration application from research to manufacturing with recirculation up to 5000L/hr.
Image of an Avantor inline dilution system
Dilution and Conditioning on Demand
Design inline dilution, conditioning or mixing systems for on-demand buffer or media preparation. Suitable for upstream and downstream processes with integrated controls.
Image of an Avantor bioprocess system sitting on pump skids
Automated Pumping and Formulation
Design pump skids with integrated software and support for advanced control fluid transfer. Compatible with your choice of pump head such as peristaltic, diaphragm, or centrifugal.
A scientist adjusting tubing on an Avantor custom bioprocess system
Expertly Engineered to Your Specifications
Avantor custom bioprocess systems are designed using a structured process that begins with your requirements and ends with a fully integrated validated solution. Our global team manages everything from component selection and chassis design to documentation and integrated systems support. Whether you need a turnkey setup or a specialty system, we work closely with your team to deliver performance, compliance, and scalability.
Trusted Process, Proven Outcomes
Icon of a person with a checklist
Experience You Can Rely On
Avantor has delivered over 850 systems worldwide, supporting small-scale builds to complex commercial installations. Backed by our cGMP manufacturing network, innovation centers, and comprehensive supply capabilities.
Icon of a stylized globe with the letters
Certified Quality and Documentation
All systems are built to ISO 9001 standards, with full support for FAT/SAT, user manuals, optional IQ/OQ documentation, and CFR Part 11 compliant software, GAMP 5, and ISA88.
Icon of a person wearing a hardhat and a sheet of paper with some test tubes
Dedicated Engineering Support
A dedicated project manager oversees your system from design to post-delivery, working closely with your team to ensure performance and compliance.
Stylized icon showing a hexagon with a cloud upload system in the center
Flexible Software Integration
Open architecture software connects upstream and downstream systems, supports SCADA, integrates with customer platforms, and is CFR Part 11-compliant.
A scientist working on a customized Avantor bioprocess system
From concept to commissioning, our global team of engineers is ready to help design a solution tailored to your bioprocess. Whether you’re scaling up or optimizing an existing line, connect with us to get started.
Consult with our Experts
Featured Resources
A scientist working on a customized Avantor bioprocess system
Explore Avantor’s full portfolio of single-use (SU) bioprocessing systems, mixing, filtration, and fluid management solutions designed to scale with your process.
Download Brochure
Image of an Avantor tangential flow filtration system
Learn how Avantor’s partnership helped overcome TFF scale-up challenges with a custom single-use system, improving yield, scalability, and compliance.
Read Case Study
Scientists in full protective gear in a buffer management facility
Find out how a hybrid buffer strategy model can help your team modernize buffer preparation to improve operational performance.
Read the White Paper