ContactUsLinkComponent
Masterflex® Peristaltic Pumps
Avantor® offers Masterflex peristaltic pumps designed for critical fluid handling workflows with a wild range of features, components and flow rates to put you in control. Engineered for long life, Masterflex provides superior accuracy and repeatability, ensuring optimal performance from the research phase to scaled-up production.
Complete pump systems for accurate fluid delivery. Preconfigured for convenience and quick set up.
An extensive offering of peristaltic pump heads that match your application and minimize downtime with faster tubing changes.
A large selection of powerful, low-maintenance drives for lab and process applications that are highly-versatile and cost-effective.
Work with our experienced engineers to design or modify a pump to meet your unique or specialized fluid handling application.
Custom extruded to fit Masterflex pumps and engineered for long life in peristaltic pump applications.
Masterflex Peristaltic Pump Series — Shop by flow range
mflax-pump-series-ISMATEC.jpg
Microflow, high accuracy
Flow Range: 0.0001 to 365 mL/min
Microflow pumps
mflx-pump-series-LS.jpg
Precise flow control
Flow Range: 0.00005 to 3400 mL/min
Precision flow
IP_series_140x140_NEW.jpg
Powerful systems
Flow Range: 0.001 to 26 LPM
Industrial flow
mflx-pump-series-BT.jpg
Highest flow rates
Flow Range: 0.3 to 42 LPM 
Highest flow pumps
How to Select the Proper Pump
Guides to help you select the best pump type and proper component configuration for your application.
How to choose the Right Masterflex Pump System