ContactUsLinkComponent
borosilicate glass D 263® T eco, thickness No. 5 (0.55±0.05 mm), CE marked 1 * 1.000 items
Catalog # MARI0106990091
Supplier:  MARIENFELD
New Product
borosilicate glass D 263® T eco, thickness No. 5 (0.55±0.05 mm), CE marked 1 * 1.000 items
Catalog # MARI0106990091
Supplier:  MARIENFELD